Assay Details
| Gene Symbol: Mplkip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025479.5 | NP_079755.1 | 2 | 2563 | 91 | |
| GenBank mRNA | AK021189.1 | - | 1 | 983 | 91 | |
| AK030630.1 | - | 2 | 2563 | 91 | ||
| AK041757.1 | - | 1 | 1465 | 91 | ||
| AK141333.1 | - | 3566 | 91 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28853136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28853134 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138816 | BC139338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138816 | BC139341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138816 | AK141333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138816 | BC117740.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138816 | AK030630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111301 | AK141333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111301 | BC117740.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111301 | AK030630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111301 | AK011919.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111301 | AK012770.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK041757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK141333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK021189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK030630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | NM_025479.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK041757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK141333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK021189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | AK030630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P322535 | NM_025479.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | AK031041.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | AK142282.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | AK146566.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | AK164135.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | BC019933.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | BC047122.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | BC052456.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | BC053718.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529169 | NM_172591.2 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128594 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138816 | BC139338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138816 | BC139341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138816 | AK141333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138816 | BC117740.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138816 | AK030630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK041757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK141333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK021189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK030630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | NM_025479.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK041757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK141333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK021189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | AK030630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P322535 | NM_025479.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111301 | AK141333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111301 | BC117740.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111301 | AK030630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111301 | AK011919.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111301 | AK012770.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28853139 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128594 | - | - |
| Whole Mouse Genome G4122A | A_52_P322535 | AK041757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | AK141333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | AK021189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | AK030630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | NM_025479.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | AK041757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | AK141333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | AK021189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | AK030630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322535 | NM_025479.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293706 | AK141333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293706 | BC117740.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293706 | AK030630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293706 | AK011919.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P293706 | AK012770.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111301 | AK141333.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111301 | BC117740.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111301 | AK030630.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111301 | AK011919.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111301 | AK012770.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128594 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28853136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28853134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28853180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10786366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10980681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10697371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10741940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10659056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10741930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10741927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28853161 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10403753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403752 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423623_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428921_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468891 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403750 | - | - |
| Mouse Genome 430 2.0 Array | 1423623_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423623_at | - | - |
| Mouse Genome 430 2.0 Array | 1428921_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428921_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66308_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66308_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300000848.mm.1 | NM_025479.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000848.mm.1 | NM_025479.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000848.mm.1 | TC1300000120.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300000849.mm.1 | NM_025479.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000849.mm.1 | NM_025479.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000849.mm.1 | TC1300000120.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423623_at | - | - |
| Mouse Expression Set 430 | 1423623_PM_at | - | - |
| Murine Genome U74v2 Set | 169753_PM_at | - | - |
| Murine Genome U74v2 Set | 169753_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423623_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423623_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423623_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423623_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428921_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428921_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 412700 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5550039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253355 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253355 | - | - |
| mouse-6 v1.1 (Illumina) | scl45038.2_639-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl45038.2_639-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1253355 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253355 | - | - |

Mplkip
Genomic Map