Assay Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009483.1 | NP_033509.1 | 7 - 8 | 960 | 69 | |
| GenBank mRNA | AJ002730.1 | - | 6 - 7 | 419 | 69 | |
| AK080370.1 | - | 7 - 8 | 979 | 69 | ||
| AK164941.1 | - | 7 - 8 | 992 | 69 | ||
| BC053433.1 | - | 7 - 8 | 960 | 6516368 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm6a
Genomic Map