Assay Details
| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102446.1 | NP_001095916.1 | 4 - 5 | 455 | 96 | |
| NM_009653.3 | NP_033783.1 | 4 - 5 | 500 | 96 | ||
| GenBank mRNA | AK002642.1 | - | 4 - 5 | 471 | 96 | |
| AK077610.1 | - | 4 - 5 | 472 | 96 | ||
| AK165257.1 | - | 4 - 5 | 463 | 96 | ||
| AK165262.1 | - | 4 - 5 | 474 | 96 | ||
| BC150870.1 | - | 4 - 5 | 447 | 9087781 | 96 | |
| M15268.1 | - | 4 - 5 | 413 | 96 | ||
| M63244.1 | - | 4 - 5 | 470 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alas2
Genomic Map