Assay Details
| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102446.1 | NP_001095916.1 | 5 - 6 | 674 | 122 | |
| NM_009653.3 | NP_033783.1 | 5 - 6 | 719 | 122 | ||
| GenBank mRNA | AK002642.1 | - | 5 - 6 | 690 | 122 | |
| AK077610.1 | - | 5 - 6 | 691 | 122 | ||
| AK165257.1 | - | 5 - 6 | 682 | 122 | ||
| AK165262.1 | - | 5 - 6 | 693 | 122 | ||
| AK172094.1 | - | 5 - 6 | 683 | 122 | ||
| BC150870.1 | - | 5 - 6 | 666 | 9087781 | 122 | |
| M15268.1 | - | 5 - 6 | 632 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alas2
Genomic Map