Assay Details
| Gene Symbol: Cryz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009968.3 | NP_034098.1 | 5 - 6 | 1060 | 63 | |
| GenBank mRNA | AK098082.1 | - | 5 - 6 | 544 | 63 | |
| AK133820.1 | - | 5 - 6 | 1061 | 63 | ||
| AK134281.1 | - | 5 - 6 | 545 | 63 | ||
| BC003800.1 | - | 5 - 6 | 552 | 3492859 | 63 | |
| BC043076.1 | - | 5 - 6 | 521 | 6404996 | 63 | |
| D78646.1 | - | 2 - 3 | 87 | 63 | ||
| S70056.1 | - | 5 - 6 | 572 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cryz
Genomic Map