Assay Details
| Gene Symbol: Dapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010019.3 | NP_034149.2 | 10 - 11 | 1066 | 81 | |
| GenBank mRNA | AB018002.1 | - | 10 - 11 | 1056 | 81 | |
| AF052942.1 | - | 8 - 9 | 746 | 81 | ||
| BC022165.1 | - | 10 - 11 | 1035 | 4016911 | 81 | |
| BX527735.1 | - | 6 - 7 | 450 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dapk2
Genomic Map