Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010143.1 | NP_034273.1 | 11 - 12 | 2526 | 92 | |
| GenBank mRNA | AK144414.1 | - | 11 - 12 | 2543 | 92 | |
| AK158043.1 | - | 12 - 13 | 3465 | 92 | ||
| BC014822.1 | - | 11 - 12 | 2527 | 3673003 | 92 | |
| BC027805.1 | - | 7 - 8 | 1813 | 92 | ||
| BC053085.1 | - | 11 - 12 | 2526 | 5708827 | 92 | |
| U11493.1 | - | 9 - 10 | 1986 | 92 | ||
| Z49086.1 | - | 11 - 12 | 2384 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb3
Genomic Map