Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010143.1 | NP_034273.1 | 1 - 2 | 469 | 140 | |
| GenBank mRNA | AK144414.1 | - | 1 - 2 | 485 | 140 | |
| BC014822.1 | - | 1 - 2 | 470 | 3673003 | 140 | |
| BC053085.1 | - | 1 - 2 | 469 | 5708827 | 140 | |
| Z49086.1 | - | 1 - 2 | 327 | 140 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC014822.1, BC053085.1, NM_010143.1, Z49086.1 transcripts.
Set Membership: |
|

Ephb3
Genomic Map