Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010143.1 | NP_034273.1 | 2 - 3 | 535 | 61 | |
| GenBank mRNA | AK144414.1 | - | 2 - 3 | 551 | 61 | |
| AK158043.1 | - | 2 - 3 | 1596 | 61 | ||
| BC014822.1 | - | 2 - 3 | 536 | 3673003 | 61 | |
| BC053085.1 | - | 2 - 3 | 535 | 5708827 | 61 | |
| Z49086.1 | - | 2 - 3 | 393 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb3
Genomic Map