Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010143.1 | NP_034273.1 | 4 - 5 | 1368 | 93 | |
| GenBank mRNA | AK144414.1 | - | 4 - 5 | 1384 | 93 | |
| AK158043.1 | - | 5 - 6 | 2308 | 93 | ||
| BC014822.1 | - | 4 - 5 | 1369 | 3673003 | 93 | |
| BC027805.1 | - | 1 - 2 | 551 | 93 | ||
| BC053085.1 | - | 4 - 5 | 1368 | 5708827 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ephb3
Genomic Map