Assay Details
| Gene Symbol: Fgf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF020738.1 | - | 4 - 5 | 683 | 91 | |
| AK011712.2 | - | 4 - 5 | 782 | 91 | ||
| AK034335.1 | - | 4 - 5 | 641 | 91 | ||
| BC030485.1 | - | 5 - 6 | 605 | 5369273 | 91 | |
| CX239635.1 | - | 5 - 6 | 590 | 91 | ||
| EU234007.1 | - | 2 - 3 | 255 | 91 | ||
| U66201.1 | - | 4 - 5 | 604 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf12
Genomic Map