Assay Details
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252282.1 | NP_001239211.1 | 18 - 19 | 2529 | 58 | |
| NM_001252283.1 | NP_001239212.1 | 18 - 19 | 2517 | 58 | ||
| NM_001252287.1 | NP_001239216.1 | 17 - 18 | 2560 | 58 | ||
| NM_001252288.1 | NP_001239217.1 | 17 - 18 | 2532 | 58 | ||
| NM_010956.4 | NP_035086.2 | 17 - 18 | 2484 | 58 | ||
| GenBank mRNA | AK147289.1 | - | 17 - 18 | 2485 | 58 | |
| AK150009.1 | - | 17 - 18 | 2450 | 58 | ||
| AK169286.1 | - | 17 - 18 | 2476 | 58 | ||
| AK220536.1 | - | 18 - 19 | 2491 | 58 | ||
| BC012042.1 | - | 1 - 2 | 119 | 58 | ||
| BC013670.1 | - | 17 - 18 | 2418 | 4188988 | 58 | |
| BC025040.1 | - | 2 - 3 | 284 | 58 | ||
| BC029143.1 | - | 2 - 3 | 294 | 58 | ||
| BC031165.1 | - | 2 - 3 | 263 | 58 | ||
| BC049104.1 | - | 17 - 18 | 2485 | 6417392 | 58 | |
| BC057354.1 | - | 18 - 19 | 2445 | 6827509 | 58 | |
More Information
Set Membership: |
|

Ogdh
Genomic Map