Assay Details
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252282.1 | NP_001239211.1 | 19 - 20 | 2598 | 90 | |
| NM_001252283.1 | NP_001239212.1 | 19 - 20 | 2586 | 90 | ||
| NM_001252287.1 | NP_001239216.1 | 18 - 19 | 2629 | 90 | ||
| NM_001252288.1 | NP_001239217.1 | 18 - 19 | 2601 | 90 | ||
| NM_010956.4 | NP_035086.2 | 18 - 19 | 2553 | 90 | ||
| GenBank mRNA | AK147289.1 | - | 18 - 19 | 2554 | 90 | |
| AK150009.1 | - | 18 - 19 | 2519 | 90 | ||
| AK169286.1 | - | 18 - 19 | 2545 | 90 | ||
| AK220536.1 | - | 19 - 20 | 2560 | 90 | ||
| BC012042.1 | - | 2 - 3 | 188 | 90 | ||
| BC013670.1 | - | 18 - 19 | 2487 | 4188988 | 90 | |
| BC025040.1 | - | 3 - 4 | 353 | 90 | ||
| BC029143.1 | - | 3 - 4 | 363 | 90 | ||
| BC031165.1 | - | 3 - 4 | 332 | 90 | ||
| BC049104.1 | - | 18 - 19 | 2554 | 6417392 | 90 | |
| BC057354.1 | - | 19 - 20 | 2514 | 6827509 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogdh
Genomic Map