Assay Details
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252282.1 | NP_001239211.1 | 6 - 7 | 800 | 101 | |
| NM_001252283.1 | NP_001239212.1 | 6 - 7 | 788 | 101 | ||
| NM_001252287.1 | NP_001239216.1 | 5 - 6 | 831 | 101 | ||
| NM_001252288.1 | NP_001239217.1 | 5 - 6 | 803 | 101 | ||
| NM_010956.4 | NP_035086.2 | 5 - 6 | 755 | 101 | ||
| GenBank mRNA | AK147289.1 | - | 5 - 6 | 756 | 101 | |
| AK150009.1 | - | 5 - 6 | 721 | 101 | ||
| AK169286.1 | - | 5 - 6 | 747 | 101 | ||
| AK220536.1 | - | 6 - 7 | 762 | 101 | ||
| BC013670.1 | - | 5 - 6 | 689 | 4188988 | 101 | |
| BC049104.1 | - | 5 - 6 | 756 | 6417392 | 101 | |
| BC057354.1 | - | 6 - 7 | 716 | 6827509 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogdh
Genomic Map