Assay Details
| Gene Symbol: Rnf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011277.2 | NP_035407.1 | 3 - 4 | 358 | 94 | |
| GenBank mRNA | AK012358.2 | - | 3 - 4 | 359 | 94 | |
| AK030319.1 | - | 4 - 5 | 531 | 94 | ||
| AK090066.1 | - | 3 - 4 | 342 | 94 | ||
| AK145767.1 | - | 3 - 4 | 365 | 94 | ||
| BC020122.1 | - | 3 - 4 | 331 | 4021046 | 94 | |
| CT010297.1 | - | 2 - 3 | 251 | 94 | ||
| Y12880.1 | - | 3 - 4 | 294 | 94 | ||
More Information
Set Membership: |
|

Rnf2
Genomic Map