Assay Details
| Gene Symbol: S100a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011311.2 | NP_035441.1 | 2 - 3 | 187 | 87 | |
| GenBank mRNA | AK003065.1 | - | 2 - 3 | 193 | 87 | |
| AK160522.1 | - | 2 - 3 | 188 | 87 | ||
| BC051214.1 | - | 1 - 2 | 146 | 1383390 | 87 | |
| BC094635.1 | - | 1 - 2 | 146 | 1248264 | 87 | |
| BX526220.1 | - | 1 - 2 | 138 | 87 | ||
| D00208.1 | - | 2 - 3 | 187 | 87 | ||
| X05835.1 | - | 1 - 2 | 144 | 87 | ||
| X16190.1 | - | 1 - 2 | 146 | 87 | ||
| Z36947.1 | - | 441 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983450 | BC094635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983450 | BX526220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983450 | BC051214.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983450 | D00208.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983450 | X05835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983448 | BC094635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983448 | BX526220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983448 | BC051214.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983448 | NM_011311.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296130 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174395 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983448 | BC094635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983448 | BX526220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983448 | BC051214.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983448 | NM_011311.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983450 | BC094635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983450 | BX526220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983450 | BC051214.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983450 | D00208.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983450 | X05835.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326061 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174395 | - | - |
| Whole Mouse Genome G4122A | A_51_P105078 | D00208.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111955 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174395 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02790915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02790910 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02790909 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02790912 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02790911 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02790884 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02344089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02790903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02790912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02743546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02743548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02344091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02743539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493815 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424542_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493812 | - | - |
| Mouse Genome 430 2.0 Array | 1424542_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424542_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20198_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20198_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006299.mm.1 | NM_011311.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006299.mm.1 | NM_011311.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006299.mm.1 | TC0300000807.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006298.mm.1 | NM_011311.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006298.mm.1 | NM_011311.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006298.mm.1 | TC0300000807.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424542_at | - | - |
| Mouse Expression Set 430 | 1424542_PM_at | - | - |
| Murine Genome U74v2 Set | 99051_PM_at | - | - |
| Murine Genome U74v2 Set | 99051_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424542_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424542_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424542_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424542_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 477051 | - | - |
| ABI Mouse Genome Survey Microarray | 906238 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3780113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2731901 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2731901 | - | - |
| mouse-6 v1.1 (Illumina) | scl020198.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020198.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2731901 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2731901 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl020198.1_11-S | - | - |

S100a4
Genomic Map