Assay Details
| Gene Symbol: Map3k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011947.3 | NP_036077.1 | 4 - 5 | 291 | 80 | |
| GenBank mRNA | AK137127.1 | - | 4 - 5 | 577 | 80 | |
| AK220450.1 | - | 4 - 5 | 585 | 80 | ||
| BC023781.1 | - | 4 - 5 | 295 | 5346982 | 80 | |
| U43187.1 | - | 4 - 5 | 594 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Map3k3
Genomic Map