Assay Details
| Gene Symbol: Homer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146153.1 | NP_001139625.1 | 7 - 8 | 859 | 74 | |
| NM_011984.2 | NP_036114.2 | 7 - 8 | 859 | 74 | ||
| GenBank mRNA | AF093261.1 | - | 5 - 6 | 686 | 74 | |
| BC005773.1 | - | 7 - 8 | 3602414 | 74 | ||
| BU054773.1 | - | 5 - 6 | 563 | 74 | ||
| BU707796.1 | - | 2 - 3 | 287 | 74 | ||
| CN539202.1 | - | 5 - 6 | 557 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Homer3
Genomic Map