Assay Details
| Gene Symbol: Vat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012037.2 | NP_036167.1 | 1 - 2 | 496 | 79 | |
| GenBank mRNA | AK034795.1 | - | 1 - 2 | 498 | 79 | |
| AK145193.1 | - | 1 - 2 | 530 | 79 | ||
| AK154965.1 | - | 1 - 2 | 479 | 79 | ||
| AK155342.1 | - | 1 - 2 | 497 | 79 | ||
| AK159310.1 | - | 1 - 2 | 439 | 79 | ||
| AK159315.1 | - | 1 - 2 | 481 | 79 | ||
| AK160891.1 | - | 1 - 2 | 488 | 79 | ||
| AK170371.1 | - | 1 - 2 | 477 | 79 | ||
| BC025496.1 | - | 1 - 2 | 452 | 5320239 | 79 | |
| BC085504.1 | - | 1 - 2 | 415 | 79 | ||
| BC099678.1 | - | 1 - 2 | 478 | 6311681 | 79 | |
| X95562.1 | - | 1 - 2 | 118 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vat1
Genomic Map