Assay Details
Gene Symbol: Vat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012037.2 | NP_036167.1 | 4 - 5 | 966 | 108 | |
GenBank mRNA | AK034795.1 | - | 4 - 5 | 968 | 108 | |
AK145193.1 | - | 4 - 5 | 1000 | 108 | ||
AK154178.1 | - | 951 | 108 | |||
AK154965.1 | - | 4 - 5 | 949 | 108 | ||
AK159310.1 | - | 4 - 5 | 909 | 108 | ||
AK159315.1 | - | 4 - 5 | 951 | 108 | ||
AK160891.1 | - | 4 - 5 | 958 | 108 | ||
AK170371.1 | - | 4 - 5 | 947 | 108 | ||
AK183725.1 | - | 2 - 3 | 220 | 107 | ||
BC024586.1 | - | 2 - 3 | 195 | 108 | ||
BC025496.1 | - | 4 - 5 | 922 | 5320239 | 108 | |
BC036169.1 | - | 3 - 4 | 408 | 108 | ||
BC085504.1 | - | 4 - 5 | 885 | 108 | ||
BC099678.1 | - | 4 - 5 | 948 | 6311681 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of mitochondrial fusion
oxidation-reduction process
zinc ion binding
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity
negative regulation of mitochondrial fusion
oxidation-reduction process
zinc ion binding
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074043 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074036 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074030 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074031 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074046 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074033 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074034 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074029 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074045 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074044 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC085504.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC099678.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC024586.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC025496.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC036169.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC085504.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC099678.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC024586.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC025496.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P79639 | BC036169.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454733 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666890 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610410 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821948 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920811 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173805 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178943 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC085504.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC099678.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC024586.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC025496.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC036169.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC085504.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC099678.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC024586.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC025496.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P79639 | BC036169.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074029 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074039 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074026 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074045 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074025 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074015 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074035 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074044 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074022 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074037 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074032 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074027 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28074018 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178943 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02411745 | - | - |
Whole Mouse Genome G4122A | A_52_P79639 | BC085504.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC099678.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC024586.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC025496.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC036169.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC085504.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC099678.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC024586.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC025496.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P79639 | BC036169.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118331 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127099 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178943 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700752 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700753 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09073254 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700755 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09391852 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09073253 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700758 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074046 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074033 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074031 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074036 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074045 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074043 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074044 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074030 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074034 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28074029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28074075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09476287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09476301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09476296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09517148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09517147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09476291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09557685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09233641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09233645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09517152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28074011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09233644 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10391455 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391458 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391457 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391456 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391460 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391459 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438165_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423726_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441930_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10391454 | - | - |
Mouse Genome 430 2.0 Array | 1441930_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1423726_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1438165_x_at | - | - |
Mouse Genome 430 2.0 Array | 1441930_x_at | - | - |
Mouse Genome 430 2.0 Array | 1438165_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1423726_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26949_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26949_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100035258.mm.1 | NM_012037.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035258.mm.1 | NM_012037.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035258.mm.1 | TC1100003807.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100035257.mm.1 | NM_012037.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035257.mm.1 | NM_012037.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035257.mm.1 | TC1100003807.mm.1 | MTA TC |
Mouse Expression Set 430 | 1423726_PM_at | - | - |
Mouse Expression Set 430 | 1423726_at | - | - |
Mouse Expression Set 430 | 1438165_x_at | - | - |
Mouse Expression Set 430 | 1438165_PM_x_at | - | - |
Mouse Expression Set 430 | 1441930_PM_x_at | - | - |
Mouse Expression Set 430 | 1441930_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1423726_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1438165_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438165_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1423726_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438165_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423726_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438165_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423726_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441930_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441930_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 930564 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1050040 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1050040 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001050040 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2440530 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2440530 | - | - |
mouse-6 v1.1 (Illumina) | scl026949.1_88-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1050040 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl026949.1_88-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050040 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2440530 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2440530 | - | - |
