Assay Details
| Gene Symbol: Sema4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013659.4 | NP_038687.2 | 12 - 13 | 1763 | 65 | |
| GenBank mRNA | AK028896.1 | - | 12 - 13 | 1726 | 65 | |
| AK030331.1 | - | 11 - 12 | 1763 | 65 | ||
| AK134973.1 | - | 11 - 12 | 1694 | 65 | ||
| AK145570.1 | - | 12 - 13 | 1764 | 65 | ||
| AK154969.1 | - | 11 - 12 | 1544 | 65 | ||
| AK173249.1 | - | 12 - 13 | 1711 | 65 | ||
| DQ073331.1 | - | 11 - 12 | 1519 | 65 | ||
| X85992.1 | - | 10 - 11 | 1355 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4b
Genomic Map