Assay Details
| Gene Symbol: Sema4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013659.4 | NP_038687.2 | 3 - 4 | 573 | 84 | |
| GenBank mRNA | AK028896.1 | - | 3 - 4 | 536 | 84 | |
| AK030331.1 | - | 2 - 3 | 573 | 84 | ||
| AK134973.1 | - | 2 - 3 | 501 | 84 | ||
| AK145570.1 | - | 3 - 4 | 574 | 84 | ||
| AK154969.1 | - | 2 - 3 | 354 | 84 | ||
| AK156620.1 | - | 2 - 3 | 407 | 84 | ||
| AK173249.1 | - | 3 - 4 | 521 | 84 | ||
| DQ073331.1 | - | 2 - 3 | 329 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4b
Genomic Map