Assay Details
| Gene Symbol: Sema4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013659.4 | NP_038687.2 | 9 - 10 | 1289 | 103 | |
| GenBank mRNA | AK028896.1 | - | 9 - 10 | 1252 | 103 | |
| AK030331.1 | - | 8 - 9 | 1289 | 103 | ||
| AK134973.1 | - | 8 - 9 | 1220 | 103 | ||
| AK145570.1 | - | 9 - 10 | 1290 | 103 | ||
| AK154969.1 | - | 8 - 9 | 1070 | 103 | ||
| AK173249.1 | - | 9 - 10 | 1237 | 103 | ||
| DQ073331.1 | - | 8 - 9 | 1045 | 103 | ||
More Information
Set Membership: |
|

Sema4b
Genomic Map