Assay Details
| Gene Symbol: Pgm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025700.2 | NP_079976.1 | 8 - 9 | 1066 | 96 | |
| GenBank mRNA | AK014332.1 | - | 8 - 9 | 991 | 96 | |
| BC030869.1 | - | 8 - 9 | 1028 | 96 | ||
| BC052762.1 | - | 8 - 9 | 1066 | 6510389 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgm1
Genomic Map