Assay Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026344.3 | NP_080620.1 | 4 - 5 | 1317 | 61 | |
| GenBank mRNA | AK011199.1 | - | 4 - 5 | 1335 | 61 | |
| AK018638.1 | - | 4 - 5 | 1318 | 61 | ||
| AK051026.1 | - | 4 - 5 | 1337 | 61 | ||
| BC014746.1 | - | 3 - 4 | 1213 | 61 | ||
| BC031124.1 | - | 4 - 5 | 1304 | 5029876 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dph2
Genomic Map