Assay Details
| Gene Symbol: Mak16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026453.3 | NP_080729.1 | 4 - 5 | 283 | 144 | |
| GenBank mRNA | AK015369.1 | - | 4 - 5 | 324 | 144 | |
| AK075982.1 | - | 4 - 5 | 284 | 144 | ||
| AK075983.1 | - | 4 - 5 | 284 | 144 | ||
| BC013079.1 | - | 4 - 5 | 298 | 3499754 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mak16
Genomic Map