Assay Details
| Gene Symbol: Pdcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026850.4 | NP_081126.2 | 3 - 4 | 259 | 73 | |
| GenBank mRNA | AF110512.1 | - | 2 - 3 | 229 | 73 | |
| AK004334.1 | - | 3 - 4 | 254 | 73 | ||
| AK078372.1 | - | 3 - 4 | 260 | 73 | ||
| AK168509.1 | - | 3 - 4 | 300 | 73 | ||
| BC005601.1 | - | 3 - 4 | 280 | 3483084 | 73 | |
More Information
Set Membership: |
|

Pdcl3
Genomic Map