| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600004024.mm.1 |
NM_027355.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004024.mm.1 |
NM_027355.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004024.mm.1 |
TC1600000436.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004026.mm.1 |
NM_027355.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004026.mm.1 |
NM_027355.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004026.mm.1 |
TC1600000436.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004023.mm.1 |
NM_027355.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004023.mm.1 |
NM_027355.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004023.mm.1 |
TC1600000436.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004025.mm.1 |
NM_027355.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004025.mm.1 |
NM_027355.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004025.mm.1 |
TC1600000436.mm.1 |
MTA TC |