Assay Details
| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027355.2 | NP_081631.2 | 2 - 3 | 1034 | 66 | |
| GenBank mRNA | AK013946.1 | - | 2 - 3 | 994 | 66 | |
| AK053769.1 | - | 2 - 3 | 1037 | 66 | ||
| BC046815.1 | - | 2 - 3 | 1006 | 5702430 | 66 | |
More Information
Set Membership: |
|

Rnf168
Genomic Map