Assay Details
| Gene Symbol: Aars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146217.4 | NP_666329.2 | 5 - 6 | 1116 | 89 | |
| GenBank mRNA | AK044451.1 | - | 5 - 6 | 867 | 89 | |
| AK085725.1 | - | 5 - 6 | 1057 | 89 | ||
| AK150227.1 | - | 5 - 6 | 1044 | 89 | ||
| AK158139.1 | - | 5 - 6 | 1065 | 89 | ||
| AY223875.1 | - | 5 - 6 | 1056 | 89 | ||
| BC013261.1 | - | 2 - 3 | 341 | 89 | ||
| BC033273.1 | - | 991 | 4196789 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aars
Genomic Map