Assay Details
| Gene Symbol: Mamdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174857.3 | NP_777282.1 | 6 - 7 | 1163 | 72 | |
| GenBank mRNA | AK035566.1 | - | 6 - 7 | 1164 | 72 | |
| AK154219.1 | - | 6 - 7 | 1240 | 72 | ||
| BC042773.1 | - | 6 - 7 | 1192 | 4019075 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mamdc2
Genomic Map