Assay Details
| Gene Symbol: Mamdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174857.3 | NP_777282.1 | 7 - 8 | 1258 | 92 | |
| GenBank mRNA | AK004794.1 | - | 6 - 7 | 1121 | 92 | |
| AK035566.1 | - | 7 - 8 | 1259 | 92 | ||
| AK154219.1 | - | 7 - 8 | 1335 | 92 | ||
| BC042773.1 | - | 7 - 8 | 1287 | 4019075 | 92 | |
More Information
Set Membership: |
|

Mamdc2
Genomic Map