Assay Details
| Gene Symbol: Mamdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174857.3 | NP_777282.1 | 9 - 10 | 1671 | 87 | |
| GenBank mRNA | AK004794.1 | - | 8 - 9 | 1534 | 87 | |
| AK035566.1 | - | 9 - 10 | 1672 | 87 | ||
| AK154219.1 | - | 9 - 10 | 1748 | 87 | ||
| BC042773.1 | - | 9 - 10 | 1700 | 4019075 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mamdc2
Genomic Map