Assay Details
| Gene Symbol: Klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174865.1 | NP_777354.1 | 4 - 5 | 610 | 102 | |
| GenBank mRNA | BC106965.1 | - | 4 - 5 | 610 | 40039744 | 102 |
| BC106966.2 | - | 4 - 5 | 607 | 102 | ||
| BC107012.1 | - | 4 - 5 | 610 | 40047687 | 102 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084945 | BC106965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084945 | BC106966.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084945 | NM_174865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084945 | BC107012.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479707 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117456 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084945 | BC106965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084945 | BC106966.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084945 | NM_174865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084945 | BC107012.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084788 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117456 | - | - |
| Whole Mouse Genome G4122A | A_52_P117282 | BC106965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117282 | BC106966.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117282 | NM_174865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117282 | BC107012.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115491 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117456 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06098612 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06098614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06142564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06339088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05989989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06339433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05989997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06475996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06036482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05989995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06284348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06034660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05904416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06230095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06476000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06367787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06339087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06539303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05989990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06034662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084809 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552544 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552542 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552545 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552540 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 685830 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6350086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6350086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2726478 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2726478 | - | - |
| mouse-6 v1.1 (Illumina) | scl0317652.3_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0317652.3_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350086 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2726478 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2726478 | - | - |

Klk15
Genomic Map