Assay Details
| Gene Symbol: Atxn2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183020.1 | NP_898841.1 | 15 - 16 | 2142 | 69 | |
| GenBank mRNA | AK155062.1 | - | 15 - 16 | 2189 | 69 | |
| AK158626.1 | - | 15 - 16 | 1829 | 69 | ||
| AK168745.1 | - | 15 - 16 | 1859 | 69 | ||
| BC034083.1 | - | 9 - 10 | 1221 | 69 | ||
| BC043451.1 | - | 10 - 11 | 1300 | 69 | ||
| BC054483.1 | - | 15 - 16 | 2142 | 6391591 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atxn2l
Genomic Map