Assay Details
| Gene Symbol: Atxn2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183020.1 | NP_898841.1 | 5 - 6 | 724 | 81 | |
| GenBank mRNA | AK084800.1 | - | 5 - 6 | 796 | 81 | |
| AK155062.1 | - | 5 - 6 | 789 | 81 | ||
| AK158626.1 | - | 5 - 6 | 411 | 81 | ||
| AK168745.1 | - | 5 - 6 | 441 | 81 | ||
| BC054483.1 | - | 5 - 6 | 724 | 6391591 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atxn2l
Genomic Map