Assay Details
| Gene Symbol: 4932414N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183113.3 | NP_898936.1 | 20 - 21 | 2024 | 143 | |
| GenBank mRNA | AK016523.1 | - | 20 - 21 | 2025 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

4932414N04Rik
Genomic Map