| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200007747.mm.1 |
NM_183113.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007747.mm.1 |
NM_183113.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007747.mm.1 |
TC0200000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200007748.mm.1 |
NM_183113.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007748.mm.1 |
NM_183113.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007748.mm.1 |
TC0200000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200007750.mm.1 |
NM_183113.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007750.mm.1 |
NM_183113.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007750.mm.1 |
TC0200000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200007751.mm.1 |
NM_183113.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007751.mm.1 |
NM_183113.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200007751.mm.1 |
TC0200000990.mm.1 |
MTA TC |