Assay Details
| Gene Symbol: Inpp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127363.1 | NP_001120835.1 | 6 - 7 | 748 | 77 | |
| NM_183144.3 | NP_898967.2 | 6 - 7 | 748 | 77 | ||
| GenBank mRNA | AK083449.1 | - | 7 - 8 | 747 | 77 | |
| AK157558.1 | - | 6 - 7 | 611 | 77 | ||
| AK171360.1 | - | 6 - 7 | 748 | 77 | ||
| AK171880.1 | - | 6 - 7 | 724 | 77 | ||
| BC056341.1 | - | 6 - 7 | 748 | 5703345 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5a
Genomic Map