Assay Details
| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183224.3 | NP_899047.1 | 3 - 4 | 589 | 65 | |
| GenBank mRNA | AK078681.1 | - | 3 - 4 | 590 | 65 | |
| AY325122.1 | - | 2 - 3 | 191 | 65 | ||
| BC118531.1 | - | 3 - 4 | 279 | 40095917 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY325122.1, BC118531.1, NM_183224.3 transcripts.
Set Membership: |
|

Fam19a3
Genomic Map