Assay Details
| Gene Symbol: Ccdc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289809.1 | NP_001276738.1 | 1 - 2 | 173 | 74 | |
| NM_001289811.1 | NP_001276740.1 | 1 - 2 | 173 | 74 | ||
| NM_183307.4 | NP_899130.1 | 2 - 3 | 372 | 74 | ||
| GenBank mRNA | AK029513.1 | - | 2 - 3 | 374 | 74 | |
| BB614558.1 | - | 2 - 3 | 373 | 74 | ||
| BC118944.1 | - | 1 - 2 | 95 | 74 | ||
| BC118945.1 | - | 1 - 2 | 95 | 40092360 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029513.1, BC118944.1, BC118945.1, NM_001289809.1, NM_001289811.1, NM_183307.4 transcripts.
Set Membership: |
|

Ccdc63
Genomic Map