Assay Details
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011535.3 | NP_035665.2 | 1 | 1040 | 132 | |
| NM_198052.2 | NP_932169.1 | 2 | 1236 | 132 | ||
| GenBank mRNA | AK031708.1 | - | 1 | 1557 | 132 | |
| AK034758.1 | - | 1 | 1600 | 132 | ||
| AK052453.1 | - | 1 | 1255 | 132 | ||
| AK145436.1 | - | 1 | 957 | 132 | ||
| AK157247.1 | - | 1 | 801 | 132 | ||
| AK172118.1 | - | 1 | 802 | 132 | ||
| BC096551.1 | - | 2 | 1236 | 30547736 | 132 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tbx3
Genomic Map