Assay Details
| Gene Symbol: Dsg1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010079.2 | NP_034209.2 | 15 | 4875 | 147 | |
| GenBank mRNA | AK029294.1 | - | 1 | 2531 | 147 | |
| AK037234.1 | - | 1 | 1748 | 147 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475111 | AK037234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475111 | AK029294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475111 | NM_010079.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475111 | AK037234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475111 | AK029294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475111 | NM_010079.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 70547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846832 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760733 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56135 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136341 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475111 | AK037234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475111 | AK029294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475111 | NM_010079.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475111 | AK037234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475111 | AK029294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475111 | NM_010079.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475111 | AK037234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475111 | AK029294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475111 | NM_010079.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475111 | AK037234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475111 | AK029294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475111 | NM_010079.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128641 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13624222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13876240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13843137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13750274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13969750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13710183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13705215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13823418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13876184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13823391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14062132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13950335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13616886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13649515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13823409 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10454115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454122 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458000_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454113 | - | - |
| Mouse Genome 430 2.0 Array | 1458000_at | - | - |
| Mouse Genome 430 2.0 Array | 1458000_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13510_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13510_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800001028.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001028.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001028.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001030.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001030.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001030.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001031.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001031.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001031.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001033.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001033.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001033.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001035.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001035.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001035.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001036.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001036.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001036.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001029.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001029.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001029.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001040.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001040.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001040.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001034.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001034.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001034.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001039.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001039.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001039.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001026.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001026.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001026.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001023.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001023.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001023.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001027.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001027.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001027.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001037.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001037.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001037.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001024.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001024.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001024.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001025.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001025.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001025.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001032.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001032.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001032.mm.1 | TC1800000153.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001038.mm.1 | NM_010079.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001038.mm.1 | NM_010079.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001038.mm.1 | TC1800000153.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1458000_PM_at | - | - |
| Mouse Expression Set 430 | 1458000_at | - | - |
| Murine Genome U74v2 Set | 166420_PM_at | - | - |
| Murine Genome U74v2 Set | 166420_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458000_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458000_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6370131 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106450324 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102230278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102230278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106450324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370131 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2714480 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2714480 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221150 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231243 | - | - |
| mouse-6 v1.1 (Illumina) | scl0013510.1_115-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102230278 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370131 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106450324 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013510.1_115-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106450324 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370131 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102230278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2714480 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2714480 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231243 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2552853 | - | - |

Dsg1a
Genomic Map