Assay Details
| Gene Symbol: Fate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003243.2 | 2 - 3 | 180 | 63 | ||
| GenBank mRNA | AK020359.1 | - | 2 - 3 | 181 | 63 | |
| BC115733.1 | - | 1 - 2 | 69 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020359.1 transcripts.
Set Membership: |
|

Fate1
Genomic Map