Assay Details
| Gene Symbol: Tmem59l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182991.2 | NP_892036.1 | 1 - 2 | 191 | 86 | |
| GenBank mRNA | AK005155.1 | - | 1 - 2 | 155 | 86 | |
| BC056172.1 | - | 1 - 2 | 191 | 5699902 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem59l
Genomic Map