Assay Details
| Gene Symbol: Tmem59l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182991.2 | NP_892036.1 | 4 - 5 | 584 | 67 | |
| GenBank mRNA | AK005155.1 | - | 4 - 5 | 548 | 67 | |
| BC056172.1 | - | 4 - 5 | 584 | 5699902 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem59l
Genomic Map