Assay Details
| Gene Symbol: Ssh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291190.1 | NP_001278119.1 | 11 - 12 | 1306 | 75 | |
| NM_177710.4 | NP_808378.2 | 12 - 13 | 1170 | 75 | ||
| GenBank mRNA | AB099288.1 | - | 12 - 13 | 1170 | 75 | |
| AK155232.1 | - | 11 - 12 | 1297 | 75 | ||
| BC141392.1 | - | 12 - 13 | 1164 | 9056005 | 75 | |
| BC141393.1 | - | 12 - 13 | 1164 | 9056006 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssh2
Genomic Map