Assay Details
| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176837.2 | NP_789807.1 | 10 - 11 | 1435 | 92 | |
| GenBank mRNA | AK014728.1 | - | 10 - 11 | 1383 | 92 | |
| AK078522.1 | - | 5 - 6 | 487 | 92 | ||
| AK087800.1 | - | 10 - 11 | 1413 | 92 | ||
| BC030858.1 | - | 10 - 11 | 1435 | 4194996 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap18
Genomic Map