Assay Details
| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176837.2 | NP_789807.1 | 1 - 2 | 187 | 69 | |
| GenBank mRNA | AK014728.1 | - | 1 - 2 | 140 | 69 | |
| AK087800.1 | - | 1 - 2 | 163 | 69 | ||
| BC030858.1 | - | 1 - 2 | 187 | 4194996 | 69 | |
More Information
Set Membership: |
|

Arhgap18
Genomic Map