Assay Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197940.2 | NP_922922.1 | 2 - 3 | 437 | 111 | |
| GenBank mRNA | AK144539.1 | - | 2 - 3 | 157 | 111 | |
| BC057854.1 | - | 2 - 3 | 226 | 6494304 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC057854.1, NM_197940.2 transcripts.
Set Membership: |
|

Wipf2
Genomic Map